Assay Details
Gene Symbol: MIR185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031178.1 | 1 | 38 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transport and golgi organization 2 homolog | TANGO2 |
C17H22orf25, C22orf25 |
Sense | Chr. 17 - 72949358 - 72967481 on NCBI Build ARS-UCD1.2 | |
Downstream | DGCR8 microprocessor complex subunit | DGCR8 | 21.3kb | Sense | Chr. 17 - 72976003 - 72990315 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
