Assay Details
Gene Symbol: MIR584-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031238.1 | 1 | 45 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transducin beta like 1 X-linked | TBL1X | -499.7kb | Anti-sense | Chr. X - 133338890 - 133585571 on NCBI Build ARS-UCD1.2 | |
Downstream | anosmin 1 | ANOS1 | 139.9kb | Sense | Chr. X - 134225253 - 134436096 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
