Assay Details
Gene Symbol: MIR10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031364.1 | 1 | 42 | 106 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-2886 | MIR2886 | bta-mir-2886 | -6.3kb | Sense | Chr. 19 - 37905897 - 37905971 on NCBI Build ARS-UCD1.2 |
Downstream | homeobox B4 | HOXB4 | 1.4kb | Sense | Chr. 19 - 37913845 - 37916665 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
