Assay Details
Gene Symbol: MIR148B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031370.1 | 1 | 38 | 72 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | COPI coat complex subunit zeta 1 | COPZ1 | Sense | Chr. 5 - 25722729 - 25742113 on NCBI Build ARS-UCD1.2 | ||
Downstream |
nuclear factor, erythroid 2 |
NFE2 | 36.2kb | Anti-sense | Chr. 5 - 25770431 - 25777486 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
