Assay Details
Gene Symbol: MIR2896 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_036363.1 | 1 | 27 | 56 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | retention in endoplasmic reticulum sorting receptor 1 | RER1 | -77.1kb | Anti-sense | Chr. 16 - 50442762 - 50452801 on NCBI Build ARS-UCD1.2 | |
Downstream | Fanconi anemia core complex associated protein 20 | FAAP20 | C16H1orf86 | 63.8kb | Sense | Chr. 16 - 50593781 - 50598411 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
