Assay Details
Gene Symbol: MIR6521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_107758.1 | 1 | 42 | 64 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | chemerin chemokine-like receptor 1 | CMKLR1 |
CMLKR1, Cmklr-1 |
-134.1kb | Anti-sense | Chr. 17 - 64539498 - 64581549 on NCBI Build ARS-UCD1.2 |
Downstream | piwi like RNA-mediated gene silencing 3 | PIWIL3 | 254.1kb | Sense | Chr. 17 - 64969916 - 64996531 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
