Assay Details
Gene Symbol: MIR6530 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_107803.1 | 1 | 40 | 52 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | membrane associated ring-CH-type finger 6 | MARCHF6 | MARCH6 | Sense | Chr. 20 - 62819405 - 62899708 on NCBI Build ARS-UCD1.2 | |
Downstream | carboxymethylenebutenolidase homolog | CMBL | 48.3kb | Anti-sense | Chr. 20 - 62947984 - 62973176 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
