Assay Details
Gene Symbol: MIR7860 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_107844.1 | 1 | 31 | 49 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | solute carrier family 25 member 30 | SLC25A30 | -23.7kb | Sense | Chr. 12 - 15541390 - 15565521 on NCBI Build ARS-UCD1.2 | |
Downstream | microRNA mir-2285h | MIR2285H | bta-mir-2285h | 40.6kb | Sense | Chr. 12 - 15629877 - 15629965 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
