Assay Details
Gene Symbol: SETD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | GU553088.1 | - | 5 - 6 | 683 | 94 | |
GU553089.1 | - | 5 - 6 | 683 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
