Assay Details
Gene Symbol: GPRC5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272010.1 | NP_001258939.1 | 3 - 4 | 1400 | 86 | |
GenBank mRNA | BC110241.1 | - | 2 - 3 | 1132 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
