Assay Details
Gene Symbol: OFD1Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281905.1 | NP_001268834.1 | 17 - 18 | 2573 | 96 | |
GenBank mRNA | JN193532.1 | - | 18 - 19 | 2747 | 96 | |
JN193533.1 | - | 17 - 18 | 2573 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
