Assay Details
Gene Symbol: OFD1Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281905.1 | NP_001268834.1 | 1 - 2 | 227 | 83 | |
GenBank mRNA | JN193532.1 | - | 1 - 2 | 227 | 83 | |
JN193533.1 | - | 1 - 2 | 227 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
