Assay Details
Gene Symbol: PAIP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302949.2 | NP_001289878.1 | 1 - 2 | 209 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001302949.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
