Assay Details
Gene Symbol: ENSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314005.1 | NP_001300934.1 | 1 - 2 | 188 | 88 | |
NM_174050.2 | NP_776475.2 | 1 - 2 | 188 | 88 | ||
GenBank mRNA | BC112682.1 | - | 1 - 2 | 185 | 88 | |
EV629449.1 | - | 1 - 2 | 191 | 88 | ||
EV691157.1 | - | 1 - 2 | 115 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
