Assay Details
Gene Symbol: KBTBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC151736.1 | - | 1 - 2 | 92 | 158 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC151736.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
