Assay Details
Gene Symbol: CIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC109722.1 | - | 2 - 3 | 233 | 122 | |
BC151807.1 | - | 2 - 3 | 261 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
