Assay Details
Gene Symbol: CIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC109722.1 | - | 5 - 6 | 381 | 98 | |
BC151807.1 | - | 5 - 6 | 409 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC109722.1 | - | 5 - 6 | 381 | 98 | |
BC151807.1 | - | 5 - 6 | 409 | 98 |
Set Membership: |
![]() ![]() ![]() ![]() |