Assay Details
Gene Symbol: SETD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | GU553088.1 | - | 6 - 7 | 805 | 98 | |
GU553089.1 | - | 6 - 7 | 805 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
