Assay Details
Gene Symbol: COX7B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244116.2 | NP_001231045.1 | 1 - 2 | 87 | 97 | |
GenBank mRNA | BC102670.1 | - | 1 - 2 | 88 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102670.1, NM_001244116.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
