Assay Details
Gene Symbol: txt-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058264.2 | NP_490665.2 | 2 - 3 | 432 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: txt-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058264.2 | NP_490665.2 | 2 - 3 | 432 | 65 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |