Assay Details
Gene Symbol: piki-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078128.6 | NP_510529.1 | 7 - 8 | 1150 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
