Assay Details
Gene Symbol: T08G11.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170907.6 | NP_740899.1 | 12 - 13 | 6818 | 104 | |
NM_171842.4 | NP_740900.1 | 13 - 14 | 6899 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
