Assay Details
Gene Symbol: T08G11.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170907.6 | NP_740899.1 | 14 - 15 | 7073 | 89 | |
NM_171842.4 | NP_740900.1 | 15 - 16 | 7154 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
