Assay Details
Gene Symbol: unc-77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306889.3 | NP_001293818.1 | 14 - 15 | 2410 | 80 | |
NM_001306890.3 | NP_001293819.1 | 13 - 14 | 2230 | 80 | ||
NM_001306891.3 | NP_001293820.1 | 14 - 15 | 2374 | 80 | ||
NM_001306894.3 | NP_001293823.1 | 14 - 15 | 2374 | 80 | ||
NM_001306895.3 | NP_001293824.1 | 15 - 16 | 2428 | 80 | ||
NM_001306898.3 | NP_001293827.1 | 15 - 16 | 2428 | 80 | ||
NM_171352.5 | NP_741413.2 | 14 - 15 | 2410 | 80 | ||
NM_171353.6 | NP_741414.2 | 13 - 14 | 2230 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
