Assay Details
Gene Symbol: sir-2.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059332.4 | NP_491733.2 | 2 - 3 | 283 | 75 | |
NR_155249.1 | 2 - 3 | 283 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
