Assay Details
Gene Symbol: sir-2.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059332.4 | NP_491733.2 | 4 - 5 | 741 | 102 | |
NR_155249.1 | 4 - 5 | 854 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
