Assay Details
Gene Symbol: ari-1.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059347.4 | NP_491748.1 | 2 - 3 | 209 | 72 |
Gene Symbol: ari-1.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383198.1 | 1 - 2 | 72 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383198.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
