Assay Details
Gene Symbol: ari-1.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383199.1 | 3 - 4 | 576 | 129 | ||
NM_059347.4 | NP_491748.1 | 4 - 5 | 703 | 129 |
Gene Symbol: ari-1.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383198.1 | 3 - 4 | 566 | 129 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383198.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
