Assay Details
Gene Symbol: gly-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059473.8 | NP_491874.1 | 1 - 2 | 215 | 90 | |
GenBank mRNA | AF154122.1 | - | 1 - 2 | 215 | 90 | |
AY037802.1 | - | 5 - 6 | 598 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
