Assay Details
Gene Symbol: sacy-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059561.5 | NP_491962.1 | 9 - 10 | 1338 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
