Assay Details
Gene Symbol: smg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025882.4 | NP_001021053.1 | 38 - 39 | 6605 | 115 | |
NM_001374908.2 | NP_001361976.1 | 38 | 6583 | 115 | ||
GenBank mRNA | AF149821.1 | - | 38 - 39 | 6605 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
