Assay Details
Gene Symbol: unc-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026700.2 | NP_001021871.1 | 16 - 17 | 2456 | 75 | |
NM_001026702.3 | NP_001021873.1 | 18 - 19 | 3482 | 75 | ||
NM_001026703.4 | NP_001021874.1 | 17 - 18 | 2465 | 75 | ||
NM_001263573.3 | NP_001250502.1 | 17 - 18 | 2474 | 75 | ||
NM_001306224.3 | NP_001293153.1 | 5 - 6 | 1554 | 75 | ||
NM_001330927.3 | NP_001317864.1 | 16 - 17 | 2465 | 75 | ||
NM_001330928.3 | NP_001317865.1 | 16 - 17 | 2420 | 75 | ||
NM_001330929.3 | NP_001317866.1 | 16 - 17 | 2411 | 75 | ||
NM_001330930.3 | NP_001317863.1 | 8 - 9 | 1433 | 75 | ||
NM_001330931.3 | NP_001317867.1 | 4 - 5 | 1412 | 75 | ||
NM_001380703.1 | NP_001367037.1 | 9 - 10 | 1442 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
