Assay Details
Gene Symbol: unc-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026700.2 | NP_001021871.1 | 17 - 18 | 2606 | 91 | |
NM_001026702.3 | NP_001021873.1 | 19 - 20 | 3632 | 91 | ||
NM_001026703.4 | NP_001021874.1 | 18 - 19 | 2615 | 91 | ||
NM_001263573.3 | NP_001250502.1 | 18 - 19 | 2624 | 91 | ||
NM_001306224.3 | NP_001293153.1 | 6 - 7 | 1704 | 91 | ||
NM_001330927.3 | NP_001317864.1 | 17 - 18 | 2615 | 91 | ||
NM_001330928.3 | NP_001317865.1 | 17 - 18 | 2570 | 91 | ||
NM_001330929.3 | NP_001317866.1 | 17 - 18 | 2561 | 91 | ||
NM_001330930.3 | NP_001317863.1 | 9 - 10 | 1583 | 91 | ||
NM_001330931.3 | NP_001317867.1 | 5 - 6 | 1562 | 91 | ||
NM_001380703.1 | NP_001367037.1 | 10 - 11 | 1592 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
