Assay Details
Gene Symbol: unc-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026700.2 | NP_001021871.1 | 19 - 20 | 3126 | 63 | |
NM_001026702.3 | NP_001021873.1 | 21 - 22 | 4152 | 63 | ||
NM_001026703.4 | NP_001021874.1 | 20 - 21 | 3135 | 63 | ||
NM_001263573.3 | NP_001250502.1 | 20 - 21 | 3144 | 63 | ||
NM_001306224.3 | NP_001293153.1 | 8 - 9 | 2224 | 63 | ||
NM_001330927.3 | NP_001317864.1 | 19 - 20 | 3135 | 63 | ||
NM_001330928.3 | NP_001317865.1 | 19 - 20 | 3090 | 63 | ||
NM_001330929.3 | NP_001317866.1 | 19 - 20 | 3081 | 63 | ||
NM_001330930.3 | NP_001317863.1 | 11 - 12 | 2103 | 63 | ||
NM_001330931.3 | NP_001317867.1 | 7 - 8 | 2082 | 63 | ||
NM_001380703.1 | NP_001367037.1 | 12 - 13 | 2112 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
