Assay Details
Gene Symbol: unc-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026700.2 | NP_001021871.1 | 27 - 28 | 5247 | 73 | |
NM_001026702.3 | NP_001021873.1 | 29 - 30 | 6273 | 73 | ||
NM_001026703.4 | NP_001021874.1 | 28 - 29 | 5256 | 73 | ||
NM_001263573.3 | NP_001250502.1 | 28 - 29 | 5265 | 73 | ||
NM_001306224.3 | NP_001293153.1 | 16 - 17 | 4345 | 73 | ||
NM_001330927.3 | NP_001317864.1 | 27 - 28 | 5256 | 73 | ||
NM_001330928.3 | NP_001317865.1 | 27 - 28 | 5211 | 73 | ||
NM_001330929.3 | NP_001317866.1 | 27 - 28 | 5202 | 73 | ||
NM_001330930.3 | NP_001317863.1 | 19 - 20 | 4224 | 73 | ||
NM_001330931.3 | NP_001317867.1 | 15 - 16 | 4203 | 73 | ||
NM_001380703.1 | NP_001367037.1 | 20 - 21 | 4233 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
