Assay Details
Gene Symbol: unc-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026700.2 | NP_001021871.1 | 9 - 10 | 1230 | 96 | |
NM_001026702.3 | NP_001021873.1 | 9 - 10 | 1230 | 96 | ||
NM_001026703.4 | NP_001021874.1 | 9 - 10 | 1230 | 96 | ||
NM_001263573.3 | NP_001250502.1 | 9 - 10 | 1239 | 96 | ||
NM_001330927.3 | NP_001317864.1 | 9 - 10 | 1239 | 96 | ||
NM_001330928.3 | NP_001317865.1 | 9 - 10 | 1194 | 96 | ||
NM_001330929.3 | NP_001317866.1 | 9 - 10 | 1185 | 96 | ||
NM_001330930.3 | NP_001317863.1 | 1 - 2 | 207 | 96 | ||
NM_001380703.1 | NP_001367037.1 | 1 - 2 | 207 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
