Assay Details
Gene Symbol: rme-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059821.5 | NP_492222.2 | 18 - 19 | 5419 | 141 | |
GenBank mRNA | AF372457.1 | - | 18 - 19 | 5419 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
