Assay Details
Gene Symbol: rme-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059821.5 | NP_492222.2 | 19 - 20 | 5449 | 102 | |
GenBank mRNA | AF372457.1 | - | 19 - 20 | 5449 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
