Assay Details
Gene Symbol: rme-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026224.6 | NP_001021395.1 | 8 - 9 | 2042 | 139 | |
NM_059821.5 | NP_492222.2 | 8 - 9 | 2035 | 139 | ||
GenBank mRNA | AF372457.1 | - | 8 - 9 | 2035 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
