Assay Details
Gene Symbol: gon-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129033.3 | NP_001122505.1 | 3 - 4 | 169 | 58 | |
NM_001373178.2 | NP_001359707.1 | 2 - 3 | 137 | 58 | ||
NM_059914.7 | NP_492315.4 | 3 - 4 | 210 | 58 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001373178.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
