Assay Details
Gene Symbol: gon-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129034.3 | NP_001122506.1 | 4 - 5 | 648 | 97 | |
NM_001373178.2 | NP_001359707.1 | 6 - 7 | 799 | 97 | ||
NM_059914.7 | NP_492315.4 | 7 - 8 | 872 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
