Assay Details
Gene Symbol: ppfr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026217.4 | NP_001021388.1 | 16 - 17 | 3220 | 87 | |
NM_001026218.5 | NP_001021389.1 | 15 - 16 | 2848 | 87 | ||
NM_001128984.4 | NP_001122456.2 | 18 - 19 | 3387 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
