Assay Details
Gene Symbol: ppfr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026217.4 | NP_001021388.1 | 23 - 24 | 4449 | 71 | |
NM_001026218.5 | NP_001021389.1 | 22 - 23 | 4077 | 71 | ||
NM_001026219.5 | NP_001021390.1 | 22 - 23 | 4347 | 71 | ||
NM_001128984.4 | NP_001122456.2 | 25 - 26 | 4616 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
