Assay Details
Gene Symbol: ppfr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026218.5 | NP_001021389.1 | 11 - 12 | 1787 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
