Assay Details
Gene Symbol: ppfr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026217.4 | NP_001021388.1 | 13 - 14 | 2424 | 151 | |
NM_001026218.5 | NP_001021389.1 | 12 - 13 | 2052 | 151 | ||
NM_001026219.5 | NP_001021390.1 | 13 - 14 | 2375 | 151 | ||
NM_001128984.4 | NP_001122456.2 | 15 - 16 | 2591 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
