Assay Details
Gene Symbol: ppfr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026217.4 | NP_001021388.1 | 12 - 13 | 2157 | 98 | |
NM_001026219.5 | NP_001021390.1 | 12 - 13 | 2108 | 98 | ||
NM_001128984.4 | NP_001122456.2 | 14 - 15 | 2324 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
