Assay Details
Gene Symbol: ppfr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026217.4 | NP_001021388.1 | 1 - 2 | 75 | 74 | |
NM_001026218.5 | NP_001021389.1 | 1 - 2 | 81 | 74 | ||
NM_001128984.4 | NP_001122456.2 | 3 - 4 | 242 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001026217.4, NM_001026218.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
