Assay Details
Gene Symbol: ppfr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026217.4 | NP_001021388.1 | 2 - 3 | 251 | 88 | |
NM_001026218.5 | NP_001021389.1 | 2 - 3 | 257 | 88 | ||
NM_001026219.5 | NP_001021390.1 | 2 - 3 | 202 | 88 | ||
NM_001128984.4 | NP_001122456.2 | 4 - 5 | 418 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
