Assay Details
Gene Symbol: fath-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060277.8 | NP_492678.1 | 3 - 4 | 347 | 89 |
Gene Symbol: C25A1.16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383713.1 | 3 - 4 | 353 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383713.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
