Assay Details
Gene Symbol: abt-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381367.1 | NP_001368466.1 | 22 - 23 | 3747 | 64 | |
NM_058548.7 | NP_490949.3 | 22 - 23 | 3858 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
